Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC78
- Entrez Gene ID
-
124093
- Gene Name
- coiled-coil domain containing 78
- Gene Aliases
-
C16orf25, CNM4, JFP10, hsCCDC78
- Chromosome Location
-
Chr.16: 722578 - 726880 on Build GRCh38
View Full Product Details