The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152467.3 | NP_689680.2 | 2 - 3 | 827 | 77 |
XM_011524706.2 | XP_011523008.1 | 4 - 5 | 960 | 77 | |
GenBank mRNA | AK057224.1 | - | 2 - 3 | 790 | 77 |
AK301797.1 | - | 2 - 3 | 731 | 77 | |
AK302141.1 | - | 2 - 3 | 775 | 77 | |
AY495339.1 | - | 2 - 3 | 809 | 77 | |
BC067753.1 | - | 2 - 3 | 814 | 77 |