The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170753.1 | NP_001164224.1 | 10 - 11 | 1112 | 106 |
NM_145017.2 | NP_659454.2 | 11 - 12 | 1172 | 106 | |
GenBank mRNA | AK057333.1 | - | 11 - 12 | 1165 | 106 |
BC043246.1 | - | 10 - 11 | 1110 | 106 | |
BC053995.1 | - | 10 - 11 | 1069 | 106 | |
JF442033.1 | - | 11 - 12 | 1145 | 106 |