Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTSF1
Entrez Gene ID
121355
Gene Name
gametocyte specific factor 1
Gene Aliases
FAM112B
Chromosome Location
Chr.12: 54455952 - 54473602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144594.2 NP_653195.2 2 - 3 106 74
XM_017018800.1 XP_016874289.1 2 - 3 79 74
GenBank mRNA AK057504.1 - 2 - 3 916 74
AK098819.1 - 2 - 3 109 74
BC021179.1 - 2 - 3 72 74
BC105587.1 - 2 - 3 107 74

View Full Product Details