The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000713.2 | NP_000704.1 | 1 - 2 | 202 | 107 |
GenBank mRNA | AK312137.1 | - | 1 - 2 | 129 | 107 |
BC109371.1 | - | 1 - 2 | 119 | 107 | |
BG340491.1 | - | 1 - 2 | 202 | 107 | |
D26308.1 | - | 1 - 2 | 150 | 107 | |
D32143.1 | - | 1 - 2 | 181 | 107 | |
EU794593.1 | - | 1 - 2 | 202 | 107 |