Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP11B1
Entrez Gene ID
1584
Gene Name
cytochrome P450 family 11 subfamily B member 1
Gene Aliases
CPN1, CYP11B, FHI, P450C11
Chromosome Location
Chr.8: 142872354 - 142879839 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000497.3 NP_000488.3 3 - 4 599 64
NM_001026213.1 NP_001021384.1 3 - 4 599 64
XM_011516874.2 XP_011515176.1 4 - 5 696 64
XM_011516875.1 XP_011515177.1 4 - 5 516 64
GenBank mRNA AF478474.1 - 3 - 4 599 64
BC096285.3 - 5 - 6 814 64
BC096286.3 - 3 - 4 599 64
BC096287.3 - 3 - 4 599 64
X55764.1 - 3 - 4 596 64

Gene Symbol
CYP11B2
Entrez Gene ID
1585
Gene Name
cytochrome P450 family 11 subfamily B member 2
Gene Aliases
ALDOS, CPN2, CYP11B, CYP11BL, CYPXIB2, P-450C18, P450C18, P450aldo
Chromosome Location
Chr.8: 142910559 - 142917843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000498.3 NP_000489.3 3 - 4 595 64
GenBank mRNA X54741.1 - 3 - 4 595 64

View Full Product Details