The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001818.3 | NP_001809.3 | 6 - 7 | 713 | 68 |
GenBank mRNA | AB031085.1 | - | 6 - 7 | 680 | 68 |
AB045829.1 | - | 6 - 7 | 680 | 68 | |
AK314988.1 | - | 6 - 7 | 754 | 68 | |
BC020744.1 | - | 6 - 7 | 713 | 68 | |
D26125.1 | - | 6 - 7 | 676 | 68 | |
M33375.1 | - | 6 - 7 | 678 | 68 | |
S68287.1 | - | 6 - 7 | 687 | 68 |