The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014077.3 | NP_054796.1 | 2 - 3 | 256 | 67 |
GenBank mRNA | AF151902.1 | - | 2 - 3 | 215 | 67 |
AL050157.1 | - | 2 - 3 | 234 | 67 | |
BC000639.1 | - | 2 - 3 | 234 | 67 | |
BC004447.1 | - | 1 - 2 | 86 | 67 | |
BC017286.1 | - | 2 - 3 | 217 | 67 | |
BG749750.1 | - | 1 - 2 | 383 | 67 |