The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020461.3 | NP_065194.2 | 22 - 23 | 5468 | 62 |
GenBank mRNA | AB051456.1 | - | 21 - 22 | 5097 | 62 |
AF272887.1 | - | 22 - 23 | 5061 | 62 | |
AL137665.1 | - | 14 - 15 | 3520 | 62 | |
AL832617.2 | - | 21 - 22 | 4650 | 62 | |
BC004127.2 | - | 4 - 5 | 601 | 62 | |
BC023573.2 | - | 8 - 9 | 2528 | 62 | |
BC144488.1 | - | 22 - 23 | 5036 | 62 | |
BC144489.1 | - | 21 - 22 | 5295 | 62 | |
BC150613.1 | - | 22 - 23 | 5036 | 62 | |
BC151209.1 | - | 22 - 23 | 5509 | 62 |