The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012162.3 | NP_036294.2 | 5 - 6 | 955 | 116 |
NM_024555.5 | NP_078831.4 | 5 - 6 | 937 | 116 | |
GenBank mRNA | AF370422.1 | - | 4 - 5 | 2268 | 116 |
AK026541.1 | - | 5 - 6 | 886 | 116 | |
AK223088.1 | - | 5 - 6 | 886 | 116 | |
AL713783.1 | - | 1 - 2 | 1166 | 116 | |
BC033066.1 | - | 5 - 6 | 944 | 116 |