The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030901.1 | NP_112163.1 | 1 - 1 | 456 | 75 |
GenBank mRNA | BC101587.1 | - | 1 - 1 | 553 | 75 |
BC117357.1 | - | 1 - 1 | 553 | 75 | |
X64993.1 | - | 1 - 1 | 82 | 75 |