Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX13B
Entrez Gene ID
56156
Gene Name
testis expressed 13B
Gene Aliases
TGC3B, TSGA5
Chromosome Location
Chr.X: 107980864 - 107982370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031273.2 NP_112563.1 2 - 3 555 115
GenBank mRNA AF285598.1 - 2 - 3 555 115

View Full Product Details