The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048164.2 | NP_001041629.1 | 7 - 8 | 1332 | 75 |
NM_032803.5 | NP_116192.4 | 7 - 8 | 1356 | 75 | |
XM_017029912.1 | XP_016885401.1 | 6 - 7 | 1847 | 75 | |
GenBank mRNA | AF320612.1 | - | 7 - 8 | 1307 | 75 |
AK027447.1 | - | 5 - 6 | 678 | 75 | |
AK074865.1 | - | 7 - 8 | 1327 | 75 | |
AK075014.1 | - | 7 - 8 | 1332 | 75 | |
BC033816.1 | - | 7 - 8 | 1311 | 75 |