Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP91
Entrez Gene ID
80829
Gene Name
ZFP91 zinc finger protein
Gene Aliases
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757
Chromosome Location
Chr.11: 58579114 - 58621550 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197051.1 NP_001183980.1 2 - 3 530 85
NM_053023.4 NP_444251.1 2 - 3 530 85
GenBank mRNA AB056107.1 - 2 - 3 476 85
AF310246.1 - 2 - 3 176 85
BC051743.1 - 2 - 3 533 85

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
-
Chromosome Location
Chr.11: 58579114 - 58625733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024091.1 2 - 3 530 85
GenBank mRNA AB057443.1 - 2 - 3 476 85
BG721319.1 - 2 - 3 539 85

View Full Product Details