Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRBA
Entrez Gene ID
987
Gene Name
LPS responsive beige-like anchor protein
Gene Aliases
BGL, CDC4L, CVID8, LAB300, LBA
Chromosome Location
Chr.4: 150264515 - 151015725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199282.2 NP_001186211.2 41 - 42 6734 59
NM_006726.4 NP_006717.2 42 - 43 6726 59
XM_005263372.3 XP_005263429.1 42 - 43 6912 59
XM_005263373.2 XP_005263430.1 42 - 43 6954 59
XM_005263374.3 XP_005263431.1 41 - 42 6878 59
XM_005263375.3 XP_005263432.1 41 - 42 6878 59
XM_011532434.2 XP_011530736.1 42 - 43 6911 59
XM_017008872.1 XP_016864361.1 41 - 42 6921 59
XM_017008873.1 XP_016864362.1 2 - 3 636 59
XM_017008874.1 XP_016864363.1 2 - 3 342 59
GenBank mRNA AB208838.1 - 17 - 18 2407 59
AF216648.2 - 42 - 43 6726 59
AF467287.1 - 41 - 42 6734 59
AK122920.1 - 5 - 6 710 59
CR749633.1 - 2 - 3 524 59
M83822.1 - 24 - 25 4162 59

View Full Product Details