The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195059.1 | NP_001181988.1 | 3 - 4 | 365 | 86 |
NM_025201.4 | NP_079477.2 | 4 - 5 | 515 | 86 | |
GenBank mRNA | AF318373.1 | - | 1 - 2 | 113 | 86 |
AF494534.1 | - | 4 - 5 | 464 | 86 | |
AK096203.1 | - | 4 - 5 | 483 | 86 | |
BC008744.2 | - | 3 - 4 | 326 | 86 |