Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRE
Entrez Gene ID
5791
Gene Name
protein tyrosine phosphatase, receptor type E
Gene Aliases
HPTPE, PTPE, R-PTP-EPSILON
Chromosome Location
Chr.10: 127907061 - 128085855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316676.1 NP_001303605.1 16 - 17 1841 95
NM_001316677.1 NP_001303606.1 14 - 15 1514 95
NM_001323354.1 NP_001310283.1 14 - 15 3234 95
NM_001323355.1 NP_001310284.1 14 - 15 1647 95
NM_001323356.1 NP_001310285.1 14 - 15 1641 95
NM_001323357.1 NP_001310286.1 11 - 12 1226 95
NM_006504.5 NP_006495.1 15 - 16 1670 95
NM_130435.4 NP_569119.1 12 - 13 1317 95
XM_005252691.2 XP_005252748.1 14 - 15 1514 95
XM_011539994.2 XP_011538296.1 14 - 15 1629 95
XM_011539995.1 XP_011538297.1 14 - 15 1632 95
XM_011539996.1 XP_011538298.1 14 - 15 1632 95
XM_017016467.1 XP_016871956.1 14 - 15 1632 95
XM_017016468.1 XP_016871957.1 14 - 15 1421 95
GenBank mRNA AJ315969.1 - 12 - 13 1226 95
AK290414.1 - 12 - 13 1253 95
AK291828.1 - 14 - 15 1514 95
AK300613.1 - 13 - 14 1407 95
BC050062.1 - 16 - 17 1749 95
X54134.1 - 15 - 16 1442 95

View Full Product Details