The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167928.1 | NP_001161400.1 | 7 - 8 | 1124 | 113 |
NM_001167929.1 | NP_001161401.1 | 5 - 6 | 873 | 113 | |
NM_001167930.1 | NP_001161402.1 | 5 - 6 | 873 | 113 | |
NM_001167931.1 | NP_001161403.1 | 6 - 7 | 960 | 113 | |
NM_002182.3 | NP_002173.1 | 6 - 7 | 960 | 113 | |
NM_134470.3 | NP_608273.1 | 6 - 7 | 960 | 113 | |
XM_017006347.1 | XP_016861836.1 | 5 - 6 | 873 | 113 | |
XM_017006348.1 | XP_016861837.1 | 2 - 3 | 274 | 113 | |
GenBank mRNA | AB006537.1 | - | 6 - 7 | 909 | 113 |
AF029213.1 | - | 4 - 5 | 712 | 113 | |
AF167343.1 | - | 4 - 5 | 703 | 113 | |
AF538730.1 | - | 6 - 7 | 909 | 113 | |
AF538731.1 | - | 5 - 6 | 822 | 113 | |
AF538732.1 | - | 5 - 6 | 822 | 113 | |
AF538733.1 | - | 5 - 6 | 822 | 113 | |
AF538734.1 | - | 6 - 7 | 909 | 113 | |
AK291669.1 | - | 7 - 8 | 1119 | 113 | |
BC053621.1 | - | 6 - 7 | 913 | 113 | |
BM760155.1 | - | 2 - 3 | 237 | 113 | |
EF591790.1 | - | 6 - 7 | 909 | 113 | |
FJ998418.1 | - | 4 - 5 | 703 | 113 |