The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138295.3 | NP_612152.1 | 3 - 4 | 321 | 67 |
XM_017011798.1 | XP_016867287.1 | 4 - 5 | 6726 | 67 | |
GenBank mRNA | AB061683.1 | - | 3 - 4 | 287 | 67 |
CN283529.1 | - | 3 - 4 | 321 | 67 |