The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127218.2 | NP_001120690.1 | 2 - 3 | 504 | 79 |
NM_001256879.1 | NP_001243808.1 | 3 - 4 | 865 | 79 | |
NM_006230.3 | NP_006221.2 | 2 - 3 | 331 | 79 | |
XM_011515431.1 | XP_011513733.1 | 2 - 3 | 327 | 79 | |
GenBank mRNA | AK292347.1 | - | 3 - 4 | 865 | 79 |
AK312294.1 | - | 2 - 3 | 464 | 79 | |
AU117477.2 | - | 2 - 3 | 309 | 79 | |
BC000459.2 | - | 2 - 3 | 294 | 79 | |
BC030001.2 | - | 1 - 2 | 271 | 79 | |
BP224472.1 | - | 2 - 3 | 331 | 79 | |
U21090.1 | - | 2 - 3 | 293 | 79 |