The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032364.5 | NP_115740.5 | 3 - 4 | 1722 | 63 |
GenBank mRNA | AF351784.1 | - | 1 - 2 | 53 | 63 |
AK055945.1 | - | 2 - 3 | 760 | 63 | |
AK123025.1 | - | 2 - 3 | 1302 | 63 | |
AK291222.1 | - | 3 - 4 | 1957 | 63 | |
AY188447.1 | - | 3 - 4 | 1639 | 63 | |
BC080655.1 | - | 2 - 3 | 674 | 63 | |
BC117146.1 | - | 3 - 4 | 1593 | 63 | |
BC117148.1 | - | 3 - 4 | 1593 | 63 | |
EF560741.1 | - | 3 - 4 | 1644 | 63 |