Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP7
Entrez Gene ID
199713
Gene Name
NLR family pyrin domain containing 7
Gene Aliases
CLR19.4, HYDM, NALP7, NOD12, PAN7, PYPAF3
Chromosome Location
Chr.19: 54923509 - 54965184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127255.1 NP_001120727.1 5 - 6 2197 69
NM_139176.3 NP_631915.2 5 - 6 2113 69
NM_206828.3 NP_996611.2 5 - 6 2197 69
XM_006723075.3 XP_006723138.1 6 - 7 2432 69
XM_006723076.3 XP_006723139.1 5 - 6 2395 69
XM_011526596.2 XP_011524898.1 7 - 8 2639 69
XM_011526599.2 XP_011524901.1 7 - 8 2650 69
XM_011526601.2 XP_011524903.1 7 - 8 2639 69
GenBank mRNA AF464765.1 - 4 - 5 2167 69
AK298630.1 - 4 - 5 1515 69
AK302655.1 - 7 - 8 2608 69
AK313964.1 - 7 - 8 2592 69
AY154462.1 - 5 - 6 2191 69
BC109124.2 - 5 - 6 2189 69
BC109125.1 - 5 - 6 2189 69

View Full Product Details