Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A6E
Entrez Gene ID
245802
Gene Name
membrane spanning 4-domains A6E
Gene Aliases
-
Chromosome Location
Chr.11: 60334882 - 60340968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139249.2 NP_640342.1 2 - 3 368 139
GenBank mRNA AF333758.1 - 2 - 3 419 139
AF354931.1 - 2 - 3 368 139
AF354932.1 - 2 - 3 252 139
AF354933.1 - 2 - 3 308 139
BC034588.1 - 2 - 3 282 139
BC069322.1 - 2 - 3 368 139
BC101783.1 - 2 - 3 428 139

View Full Product Details