The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005557.3 | NP_005548.2 | 6 - 7 | 1422 | 89 |
GenBank mRNA | AF061812.1 | - | 6 - 7 | 1297 | 89 |
AK290853.1 | - | 6 - 7 | 1360 | 89 | |
BC039169.1 | - | 6 - 7 | 1333 | 89 | |
S79867.1 | - | 6 - 7 | 1283 | 89 |