The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021615.4 | NP_067628.1 | 1 - 2 | 87 | 102 |
GenBank mRNA | AF280086.1 | - | 1 - 2 | 95 | 102 |