The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021810.4 | NP_068582.2 | 5 - 6 | 503 | 68 |
NM_177980.2 | NP_817089.1 | 17 - 18 | 2606 | 68 | |
XM_017027998.1 | XP_016883487.1 | 17 - 18 | 3273 | 68 | |
XM_017027999.1 | XP_016883488.1 | 16 - 17 | 3204 | 68 | |
XM_017028001.1 | XP_016883490.1 | 15 - 16 | 3150 | 68 | |
GenBank mRNA | AK055202.1 | - | 17 - 18 | 2595 | 68 |
AK074477.1 | - | 5 - 6 | 491 | 68 | |
BC062570.1 | - | 3 - 4 | 380 | 68 | |
BC136306.1 | - | 17 - 18 | 2589 | 68 | |
BC143931.1 | - | 17 - 18 | 2589 | 68 |