The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145243.4 | NP_660286.1 | 2 - 3 | 613 | 89 |
GenBank mRNA | AB048348.1 | - | 2 - 3 | 596 | 89 |
AK308847.1 | - | 2 - 3 | 611 | 89 | |
BC012915.1 | - | 2 - 3 | 594 | 89 | |
BU191827.1 | - | 2 - 3 | 586 | 89 |