Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASE11
Entrez Gene ID
122651
Gene Name
ribonuclease A family member 11 (inactive)
Gene Aliases
C14orf6, HEL-S-84p, RAJ1
Chromosome Location
Chr.14: 20582892 - 20590258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145250.3 NP_660293.1 2 - 3 160 84
GenBank mRNA AK292348.1 - 1 - 2 161 84
AK314065.1 - 2 - 3 312 84
AY358794.1 - 2 - 3 440 84
BC025410.1 - 2 - 3 87 84
BX161458.1 - 2 - 3 132 84
CB995247.1 - 2 - 3 186 84
FJ237361.1 - 2 - 3 87 84

View Full Product Details