The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000794.3 | NP_000785.1 | 1 - 2 | 462 | 110 |
GenBank mRNA | DA249160.1 | - | 1 - 2 | 484 | 110 |