Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO15
Entrez Gene ID
201456
Gene Name
F-box protein 15
Gene Aliases
FBX15
Chromosome Location
Chr.18: 74073343 - 74147865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152676.2 NP_689889.1 1 - 2 230 59
GenBank mRNA BC029579.1 - 1 - 2 180 59
BI829755.1 - 1 - 2 58

View Full Product Details