The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138492.5 | NP_612501.3 | 4 - 5 | 532 | 89 |
NM_205846.2 | NP_995318.1 | 3 - 4 | 329 | 89 | |
XM_017009135.1 | XP_016864624.1 | 3 - 4 | 329 | 89 | |
GenBank mRNA | AK125215.1 | - | 3 - 4 | 264 | 89 |
BC012033.1 | - | 4 - 5 | 499 | 89 | |
BC028175.1 | - | - | 246 | 89 | |
BQ645893.1 | - | 3 - 4 | 299 | 89 | |
BU167299.1 | - | 3 - 4 | 191 | 89 |