Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 19 - 20 3904 78
NM_001144759.2 NP_001138231.1 18 - 19 3763 78
NM_015157.3 NP_055972.1 20 - 21 4127 78
XM_005271468.3 XP_005271525.1 20 - 21 4718 78
XM_005271469.3 XP_005271526.1 20 - 21 4718 78
XM_005271470.3 XP_005271527.1 19 - 20 4673 78
XM_005271471.3 XP_005271528.1 19 - 20 4673 78
XM_005271472.3 XP_005271529.1 19 - 20 4577 78
XM_005271473.3 XP_005271530.1 18 - 19 4532 78
XM_005271474.4 XP_005271531.1 18 - 19 4532 78
XM_005277680.3 XP_005277737.1 23 - 24 5159 78
XM_006718796.3 XP_006718859.1 23 - 24 4348 78
XM_006718797.3 XP_006718860.1 22 - 23 5018 78
XM_006718798.3 XP_006718861.1 21 - 22 4847 78
XM_006718799.3 XP_006718862.1 20 - 21 4802 78
XM_006718800.3 XP_006718863.1 20 - 21 4802 78
XM_006718801.3 XP_006718864.1 20 - 21 4706 78
XM_006718802.3 XP_006718865.1 19 - 20 4661 78
XM_006718803.3 XP_006718866.1 19 - 20 4661 78
XM_011542703.2 XP_011541005.1 23 - 24 5162 78
XM_011542704.2 XP_011541006.1 23 - 24 5162 78
XM_011542705.2 XP_011541007.1 22 - 23 5117 78
XM_011542706.2 XP_011541008.1 22 - 23 5117 78
XM_011542707.2 XP_011541009.1 22 - 23 5033 78
XM_011542708.2 XP_011541010.1 23 - 24 4351 78
XM_011542709.2 XP_011541011.1 23 - 24 4661 78
XM_011542710.2 XP_011541012.1 22 - 23 4991 78
XM_011542711.2 XP_011541013.1 20 - 21 4068 78
XM_011542713.2 XP_011541015.1 20 - 21 4062 78
XM_011542715.2 XP_011541017.1 13 - 14 3039 78
XM_017017407.1 XP_016872896.1 22 - 23 5030 78
XM_017017408.1 XP_016872897.1 21 - 22 4973 78
XM_017017409.1 XP_016872898.1 21 - 22 4946 78
XM_017017410.1 XP_016872899.1 21 - 22 4889 78
GenBank mRNA AB014538.2 - 19 - 20 3881 78
AB094090.1 - 20 - 21 4127 78
AB177859.1 - 15 - 16 3053 78
AK027510.1 - 10 - 11 1392 78
AK074070.2 - 18 - 19 3712 78
AK074747.1 - 4 - 5 647 78
AK074918.1 - 12 - 13 1599 78
AK125783.1 - 21 - 22 3131 78
AL832373.1 - 17 - 18 3255 78
BC013031.2 - 12 - 13 1749 78
BC014510.1 - 5 - 6 410 78
BC098586.1 - 18 - 19 4257 78
BX648176.1 - 13 - 14 1915 78
CR749783.1 - 19 - 20 4420 78

View Full Product Details