Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127454306-127454819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001005374.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1701753 c.43C>T p.R15W Chr.9: 127454570-127454570 on Build GRCh38
4546915 c.40G>T p.A14S Chr.9: 127454567-127454567 on Build GRCh38
6704602 c.44G>A p.R15Q Chr.9: 127454571-127454571 on Build GRCh38
6342928 c.57G>T p.E19D Chr.9: 127454584-127454584 on Build GRCh38
6229932 c.4C>T p.P2S Chr.9: 127454531-127454531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998971851 Chr.9: 127454387-127454387 on Build GRCh38
rs201473713 Chr.9: 127454540-127454540 on Build GRCh38
rs924073658 Chr.9: 127454789-127454789 on Build GRCh38
rs2243509 Chr.9: 127454771-127454771 on Build GRCh38
rs2244218 Chr.9: 127454672-127454672 on Build GRCh38
rs772129554 Chr.9: 127454485-127454485 on Build GRCh38
rs748153917 Chr.9: 127454482-127454482 on Build GRCh38
rs372408222 Chr.9: 127454589-127454589 on Build GRCh38
rs749714585 Chr.9: 127454414-127454414 on Build GRCh38
rs1014645241 Chr.9: 127454729-127454730 on Build GRCh38
rs1021228188 Chr.9: 127454352-127454352 on Build GRCh38
rs545615822 Chr.9: 127454686-127454686 on Build GRCh38
rs1024229804 Chr.9: 127454698-127454698 on Build GRCh38
rs896366180 Chr.9: 127454337-127454337 on Build GRCh38
rs778740625 Chr.9: 127454544-127454544 on Build GRCh38
rs753421683 Chr.9: 127454620-127454620 on Build GRCh38
rs375949142 Chr.9: 127454626-127454626 on Build GRCh38
rs759751440 Chr.9: 127454510-127454510 on Build GRCh38
rs750997053 Chr.9: 127454535-127454537 on Build GRCh38
rs776039231 Chr.9: 127454577-127454577 on Build GRCh38
rs140467743 Chr.9: 127454543-127454543 on Build GRCh38
rs528797347 Chr.9: 127454788-127454788 on Build GRCh38
rs753536686 Chr.9: 127454539-127454539 on Build GRCh38
rs764897663 Chr.9: 127454628-127454628 on Build GRCh38
rs917571389 Chr.9: 127454751-127454751 on Build GRCh38
rs557444588 Chr.9: 127454412-127454412 on Build GRCh38
rs770259167 Chr.9: 127454576-127454576 on Build GRCh38
rs957360263 Chr.9: 127454426-127454426 on Build GRCh38
rs777784051 Chr.9: 127454550-127454550 on Build GRCh38
rs752596738 Chr.9: 127454549-127454549 on Build GRCh38
rs777868589 Chr.9: 127454491-127454491 on Build GRCh38
rs765893788 Chr.9: 127454537-127454537 on Build GRCh38
rs78037809 Chr.9: 127454346-127454346 on Build GRCh38
rs74876850 Chr.9: 127454644-127454644 on Build GRCh38
rs537550075 Chr.9: 127454370-127454370 on Build GRCh38
rs766700610 Chr.9: 127454598-127454599 on Build GRCh38
rs368646898 Chr.9: 127454571-127454571 on Build GRCh38
rs534992818 Chr.9: 127454336-127454336 on Build GRCh38
rs142913281 Chr.9: 127454569-127454569 on Build GRCh38
rs961780308 Chr.9: 127454739-127454739 on Build GRCh38
rs540942030 Chr.9: 127454536-127454536 on Build GRCh38
rs75663017 Chr.9: 127454710-127454710 on Build GRCh38
rs776125314 Chr.9: 127454522-127454522 on Build GRCh38
rs746999697 Chr.9: 127454555-127454555 on Build GRCh38
rs1003720087 Chr.9: 127454721-127454721 on Build GRCh38
rs1053696036 Chr.9: 127454668-127454668 on Build GRCh38
rs545820339 Chr.9: 127454747-127454747 on Build GRCh38
rs776922280 Chr.9: 127454509-127454509 on Build GRCh38
rs763601883 Chr.9: 127454530-127454530 on Build GRCh38
rs769236666 Chr.9: 127454591-127454591 on Build GRCh38
rs944532459 Chr.9: 127454786-127454786 on Build GRCh38
rs747142126 Chr.9: 127454492-127454492 on Build GRCh38
rs970915843 Chr.9: 127454440-127454440 on Build GRCh38
rs371146474 Chr.9: 127454645-127454645 on Build GRCh38
rs1023755188 Chr.9: 127454429-127454429 on Build GRCh38
rs553074777 Chr.9: 127454532-127454532 on Build GRCh38
rs919454589 Chr.9: 127454513-127454513 on Build GRCh38
rs958269450 Chr.9: 127454391-127454391 on Build GRCh38
rs771006038 Chr.9: 127454508-127454508 on Build GRCh38
rs1017867713 Chr.9: 127454464-127454464 on Build GRCh38
rs543119820 Chr.9: 127454667-127454667 on Build GRCh38
rs1012444161 Chr.9: 127454673-127454673 on Build GRCh38
rs911722051 Chr.9: 127454641-127454641 on Build GRCh38
rs902530272 Chr.9: 127454653-127454653 on Build GRCh38
rs986107067 Chr.9: 127454439-127454439 on Build GRCh38
rs771047258 Chr.9: 127454560-127454560 on Build GRCh38
rs760040162 Chr.9: 127454603-127454603 on Build GRCh38
rs935363017 Chr.9: 127454657-127454657 on Build GRCh38
rs570954827 Chr.9: 127454478-127454478 on Build GRCh38
rs765985792 Chr.9: 127454615-127454615 on Build GRCh38
rs572330792 Chr.9: 127454533-127454533 on Build GRCh38
rs1042296620 Chr.9: 127454784-127454784 on Build GRCh38
rs765806160 Chr.9: 127454751-127454752 on Build GRCh38
rs548793451 Chr.9: 127454345-127454345 on Build GRCh38
rs539859583 Chr.9: 127454515-127454515 on Build GRCh38
rs953310785 Chr.9: 127454765-127454765 on Build GRCh38
rs774936278 Chr.9: 127454594-127454594 on Build GRCh38
rs760944483 Chr.9: 127454547-127454548 on Build GRCh38
rs781265596 Chr.9: 127454570-127454570 on Build GRCh38

View Full Product Details