The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290256.1 | NP_001277185.1 | 4 - 5 | 1620 | 69 |
NM_001290257.1 | NP_001277186.1 | 4 - 5 | 1555 | 69 | |
NM_015299.2 | NP_056114.1 | 4 - 5 | 1683 | 69 | |
XM_005267474.4 | XP_005267531.1 | 4 - 5 | 1639 | 69 | |
XM_011536590.2 | XP_011534892.1 | 4 - 5 | 1789 | 69 | |
XM_017021114.1 | XP_016876603.1 | 4 - 5 | 1555 | 69 | |
GenBank mRNA | AB002321.1 | - | 4 - 5 | 1620 | 69 |
AK291680.1 | - | 4 - 5 | 1536 | 69 | |
BC015780.1 | - | 2 - 3 | 706 | 69 | |
BC042554.1 | - | 4 - 5 | 1648 | 69 | |
BX161465.1 | - | 4 - 5 | 1656 | 69 |