The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012393.2 | NP_036525.1 | 11 - 12 | 1480 | 67 |
XM_006721546.2 | XP_006721609.1 | 11 - 12 | 1503 | 67 | |
XM_017024760.1 | XP_016880249.1 | 10 - 11 | 1282 | 67 | |
GenBank mRNA | AB002359.1 | - | 11 - 12 | 1439 | 67 |
AK292402.1 | - | 11 - 12 | 1474 | 67 | |
AK292804.1 | - | 11 - 12 | 1480 | 67 | |
AK295895.1 | - | 9 - 10 | 1398 | 67 | |
BC146768.1 | - | 11 - 12 | 1439 | 67 | |
BC167158.1 | - | 11 - 12 | 1439 | 67 |