The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015267.3 | NP_056082.2 | 4 - 5 | 460 | 73 |
XM_011538061.2 | XP_011536363.1 | 4 - 5 | 1970 | 73 | |
XM_011538069.2 | XP_011536371.1 | 4 - 5 | 309 | 73 | |
XM_011538070.2 | XP_011536372.1 | 4 - 5 | 1219 | 73 | |
XM_017019080.1 | XP_016874569.1 | 5 - 6 | 1302 | 73 | |
XM_017019081.1 | XP_016874570.1 | 5 - 6 | 365 | 73 | |
GenBank mRNA | AB006631.2 | - | 4 - 5 | 459 | 73 |
AF234996.1 | - | 5 - 6 | 623 | 73 | |
AF271236.1 | - | 4 - 5 | 370 | 73 | |
BC151245.1 | - | 4 - 5 | 470 | 73 |