Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRP4
Entrez Gene ID
4038
Gene Name
LDL receptor related protein 4
Gene Aliases
CLSS, CMS17, LRP-4, LRP10, MEGF7, SOST2
Chromosome Location
Chr.11: 46854715 - 46918622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002334.3 NP_002325.2 17 - 18 2665 66
XM_011520103.2 XP_011518405.1 11 - 12 1735 66
XM_011520104.2 XP_011518406.1 2 - 3 202 66
XM_017017734.1 XP_016873223.1 17 - 18 2582 66
GenBank mRNA AB011540.1 - 9 - 10 1435 66
AB084910.1 - 17 - 18 2665 66
AB231861.1 - 17 - 18 2568 66
BC136667.1 - 17 - 18 2586 66
BC136668.1 - 17 - 18 2586 66

View Full Product Details