Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 12 - 13 2868 79
NM_015157.3 NP_055972.1 13 - 14 3091 79
XM_005271468.3 XP_005271525.1 12 - 13 3637 79
XM_005271469.3 XP_005271526.1 12 - 13 3637 79
XM_005271470.3 XP_005271527.1 12 - 13 3637 79
XM_005271471.3 XP_005271528.1 12 - 13 3637 79
XM_006718798.3 XP_006718861.1 13 - 14 3766 79
XM_006718799.3 XP_006718862.1 13 - 14 3766 79
XM_006718800.3 XP_006718863.1 13 - 14 3766 79
GenBank mRNA AB014538.2 - 12 - 13 2845 79
AB094090.1 - 13 - 14 3091 79
AB177859.1 - 7 - 8 1972 79
AK125783.1 - 14 - 15 2095 79

View Full Product Details