Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 21 - 22 2116 119
NM_145695.2 NP_663733.1 21 - 22 2116 119
XM_005249628.2 XP_005249685.1 22 - 23 2644 119
XM_005249629.2 XP_005249686.1 22 - 23 2641 119
XM_005249630.2 XP_005249687.1 21 - 22 2157 119
XM_005249631.3 XP_005249688.1 20 - 21 2372 119
XM_011515153.2 XP_011513455.1 22 - 23 2482 119
XM_011515154.2 XP_011513456.1 22 - 23 2198 119
XM_011515156.2 XP_011513458.1 21 - 22 2154 119
XM_011515157.2 XP_011513459.1 21 - 22 2161 119
XM_011515158.2 XP_011513460.1 20 - 21 2297 119
XM_017011783.1 XP_016867272.1 22 - 23 2535 119
XM_017011784.1 XP_016867273.1 22 - 23 2195 119
XM_017011785.1 XP_016867274.1 21 - 22 2429 119
XM_017011786.1 XP_016867275.1 21 - 22 2409 119
XM_017011787.1 XP_016867276.1 21 - 22 2159 119
XM_017011788.1 XP_016867277.1 21 - 22 2445 119
XM_017011789.1 XP_016867278.1 21 - 22 2162 119
XM_017011790.1 XP_016867279.1 22 - 23 2481 119
GenBank mRNA AB018261.1 - 18 - 19 1745 119
AK304719.1 - 20 - 21 2372 119
BC105005.1 - 21 - 22 1996 119
BC143632.1 - 21 - 22 1993 119
BC143633.1 - 20 - 21 1955 119

View Full Product Details