The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014014.4 | NP_054733.2 | 11 - 12 | 1514 | 66 |
XM_017003603.1 | XP_016859092.1 | 13 - 14 | 2344 | 66 | |
GenBank mRNA | AB018331.2 | - | 9 - 10 | 1052 | 66 |
AK023840.1 | - | 5 - 6 | 526 | 66 | |
AK024391.1 | - | 5 - 6 | 595 | 66 | |
AK090671.1 | - | 11 - 12 | 1491 | 66 | |
AL831994.1 | - | 2 - 3 | 230 | 66 | |
AY572488.1 | - | 11 - 12 | 1460 | 66 | |
BC065924.1 | - | 4 - 5 | 437 | 66 | |
BC112891.1 | - | 5 - 6 | 635 | 66 |