The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080421.2 | NP_001073890.2 | 23 - 24 | 2853 | 85 |
XM_011527810.2 | XP_011526112.1 | 22 - 23 | 2968 | 85 | |
XM_011527811.2 | XP_011526113.1 | 23 - 24 | 2998 | 85 | |
XM_017026502.1 | XP_016881991.1 | 23 - 24 | 2998 | 85 | |
GenBank mRNA | AB028955.2 | - | 23 - 24 | 2851 | 85 |