The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153252.4 | NP_694984.4 | 19 - 20 | 2494 | 74 |
XM_005262113.3 | XP_005262170.1 | 19 - 20 | 2496 | 74 | |
XM_017029384.1 | XP_016884873.1 | 15 - 16 | 2024 | 74 | |
XM_017029385.1 | XP_016884874.1 | 19 - 20 | 2496 | 74 | |
GenBank mRNA | AK095887.1 | - | 2 - 3 | 367 | 74 |
AY497046.1 | - | 19 - 20 | 2494 | 74 | |
AY497047.1 | - | 16 - 17 | 2426 | 74 | |
AY497048.1 | - | 19 - 20 | 2517 | 74 | |
AY497049.1 | - | 18 - 19 | 2487 | 74 | |
AY497050.1 | - | 19 - 20 | 2467 | 74 | |
AY497051.1 | - | 16 - 17 | 2054 | 74 | |
AY497052.1 | - | 21 - 22 | 2653 | 74 | |
AY497053.1 | - | 17 - 18 | 2242 | 74 | |
AY497054.1 | - | 16 - 17 | 2143 | 74 | |
AY497055.1 | - | 19 - 20 | 2428 | 74 | |
AY497056.1 | - | 16 - 17 | 2399 | 74 | |
AY497057.1 | - | 15 - 16 | 2298 | 74 | |
AY497058.1 | - | 15 - 16 | 2137 | 74 | |
AY497059.1 | - | 17 - 18 | 2508 | 74 | |
AY497060.1 | - | 13 - 14 | 2075 | 74 | |
BC111490.1 | - | 19 - 20 | 2444 | 74 |