Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OIT3
Entrez Gene ID
170392
Gene Name
oncoprotein induced transcript 3
Gene Aliases
LZP
Chromosome Location
Chr.10: 72893556 - 72933036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152635.2 NP_689848.1 8 - 9 1714 82
NR_130125.1 7 - 8 1298 82
GenBank mRNA AK096435.1 - 8 - 9 1689 82
AY013707.2 - 8 - 9 1545 82
AY358339.1 - 8 - 9 1667 82
BC126440.1 - 7 - 8 1270 82

View Full Product Details