Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
Chromosome Location
Chr.19: 15508487 - 15552317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173483.3 NP_775754.2 11 - 12 1437 73
XM_011527692.2 XP_011525994.1 12 - 13 1679 73
XM_011527693.2 XP_011525995.1 11 - 12 1449 73
GenBank mRNA AK096820.1 - 11 - 12 1437 73
BC069351.1 - 10 - 11 1374 73
BC093894.1 - 10 - 11 1311 73
BC093896.1 - 10 - 11 1311 73

View Full Product Details