Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ULK3
Entrez Gene ID
25989
Gene Name
unc-51 like kinase 3
Gene Aliases
-
Chromosome Location
Chr.15: 74836116 - 74843211 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099436.3 NP_001092906.3 12 - 13 1347 114
NM_001284364.2 NP_001271293.2 12 - 13 1347 114
NM_001284365.2 NP_001271294.1 12 - 13 1270 114
NR_104307.2 11 - 12 1528 114
XM_005254289.2 XP_005254346.1 13 - 14 1362 114
XM_017022068.1 XP_016877557.1 13 - 14 1396 114
XM_017022069.1 XP_016877558.1 13 - 14 1400 114
XM_017022070.1 XP_016877559.1 13 - 14 1413 114
XM_017022071.1 XP_016877560.1 13 - 14 1400 114
XM_017022072.1 XP_016877561.1 12 - 13 1576 114
GenBank mRNA AK293181.1 - 12 - 13 1200 114
AK294241.1 - 12 - 13 1281 114
AK294245.1 - 12 - 13 1277 114
AK294251.1 - 12 - 13 1281 114
AK298879.1 - 12 - 13 1204 114
AK299380.1 - 12 - 13 1274 114
AL117482.1 - 11 - 12 1663 114
AL360256.1 - 7 - 8 940 114
BC157884.1 - 12 - 13 1329 114

View Full Product Details