Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNRC6A
Entrez Gene ID
27327
Gene Name
trinucleotide repeat containing 6A
Gene Aliases
CAGH26, GW1, GW182, TNRC6
Chromosome Location
Chr.16: 24610260 - 24826227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014494.2 NP_055309.2 15 - 16 4365 96
XM_005255254.3 XP_005255311.1 14 - 15 4208 96
XM_005255257.4 XP_005255314.1 14 - 15 4332 96
XM_017023143.1 XP_016878632.1 14 - 15 4309 96
XM_017023144.1 XP_016878633.1 13 - 14 4276 96
XM_017023145.1 XP_016878634.1 14 - 15 4242 96
XM_017023146.1 XP_016878635.1 14 - 15 4374 96
XM_017023147.1 XP_016878636.1 14 - 15 4402 96
XM_017023148.1 XP_016878637.1 13 - 14 4129 96
XM_017023149.1 XP_016878638.1 12 - 13 4126 96
XM_017023150.1 XP_016878639.1 13 - 14 4091 96
XM_017023151.1 XP_016878640.1 12 - 13 3980 96
XM_017023152.1 XP_016878641.1 13 - 14 3881 96
XM_017023153.1 XP_016878642.1 14 - 15 4332 96
XM_017023154.1 XP_016878643.1 12 - 13 4223 96
GenBank mRNA AB040893.1 - 10 - 11 2565 96
AJ492221.1 - 14 - 15 4251 96
AK025696.1 - 8 - 9 922 96
AK094542.1 - 2 - 3 947 96
AL713750.1 - 2 - 3 1118 96
AL833757.1 - 9 - 10 1526 96
AY035864.1 - 12 - 13 3899 96

View Full Product Details