The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000260.3 | NP_000251.3 | 20 - 21 | 104 | |
NM_001127179.2 | NP_001120651.2 | 20 - 21 | 104 | ||
NM_001127180.1 | NP_001120652.1 | 20 - 21 | 104 | ||
XM_011545044.2 | XP_011543346.1 | 20 - 21 | 104 | ||
XM_011545046.2 | XP_011543348.2 | 19 - 20 | 104 | ||
XM_011545050.2 | XP_011543352.1 | 17 - 18 | 104 | ||
XM_017017778.1 | XP_016873267.1 | 19 - 20 | 104 | ||
XM_017017779.1 | XP_016873268.1 | 19 - 20 | 104 | ||
XM_017017780.1 | XP_016873269.1 | 19 - 20 | 104 | ||
XM_017017781.1 | XP_016873270.1 | 19 - 20 | 104 | ||
XM_017017782.1 | XP_016873271.1 | 19 - 20 | 104 | ||
XM_017017783.1 | XP_016873272.1 | 19 - 20 | 104 | ||
XM_017017784.1 | XP_016873273.1 | 19 - 20 | 104 | ||
XM_017017785.1 | XP_016873274.1 | 17 - 18 | 104 | ||
XM_017017786.1 | XP_016873275.1 | 19 - 20 | 104 | ||
XM_017017787.1 | XP_016873276.1 | 19 - 20 | 104 | ||
XM_017017788.1 | XP_016873277.1 | 19 - 20 | 104 | ||
GenBank mRNA | BC023212.1 | - | 5 - 6 | 104 | |
BC144519.1 | - | 21 - 22 | 104 | ||
U34227.1 | - | 20 - 21 | 2640 | 104 | |
U39226.1 | - | 20 - 21 | 104 | ||
U55208.1 | - | 20 - 21 | 2640 | 104 | |
U55209.1 | - | 20 - 21 | 2640 | 104 |