Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34231549-34232136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

SNP Detail

SNP ID Chromosome Location
rs919685905 Chr.15: 34231576-34231576 on Build GRCh38
rs894710143 Chr.15: 34231595-34231596 on Build GRCh38
rs911661329 Chr.15: 34231780-34231780 on Build GRCh38
rs1014800155 Chr.15: 34231869-34231869 on Build GRCh38
rs987520352 Chr.15: 34231774-34231774 on Build GRCh38
rs916101390 Chr.15: 34231782-34231782 on Build GRCh38
rs886051042 Chr.15: 34231661-34231661 on Build GRCh38
rs576342316 Chr.15: 34231850-34231850 on Build GRCh38
rs537801396 Chr.15: 34232022-34232022 on Build GRCh38
rs755356379 Chr.15: 34231577-34231580 on Build GRCh38
rs190767469 Chr.15: 34231994-34231994 on Build GRCh38
rs1053131287 Chr.15: 34231852-34231852 on Build GRCh38
rs1028934580 Chr.15: 34231952-34231952 on Build GRCh38
rs535022392 Chr.15: 34231752-34231752 on Build GRCh38
rs1049713817 Chr.15: 34231593-34231596 on Build GRCh38
rs1024374712 Chr.15: 34231701-34231701 on Build GRCh38
rs6495639 Chr.15: 34232035-34232035 on Build GRCh38
rs1034350536 Chr.15: 34231674-34231674 on Build GRCh38
rs887206619 Chr.15: 34231839-34231839 on Build GRCh38
rs952488971 Chr.15: 34232066-34232066 on Build GRCh38
rs977131625 Chr.15: 34232081-34232081 on Build GRCh38
rs926150990 Chr.15: 34232082-34232082 on Build GRCh38
rs988012809 Chr.15: 34231642-34231642 on Build GRCh38
rs931081060 Chr.15: 34231780-34231781 on Build GRCh38
rs951521925 Chr.15: 34231736-34231736 on Build GRCh38
rs181064062 Chr.15: 34231620-34231620 on Build GRCh38
rs931619573 Chr.15: 34231754-34231754 on Build GRCh38
rs936290190 Chr.15: 34231793-34231793 on Build GRCh38
rs113459768 Chr.15: 34231592-34231592 on Build GRCh38
rs1012108177 Chr.15: 34231874-34231874 on Build GRCh38
rs373755957 Chr.15: 34231870-34231870 on Build GRCh38
rs540719442 Chr.15: 34231596-34231596 on Build GRCh38
rs1046116385 Chr.15: 34231836-34231836 on Build GRCh38
rs1014273612 Chr.15: 34231833-34231833 on Build GRCh38
rs770260783 Chr.15: 34231592-34231593 on Build GRCh38
rs994780771 Chr.15: 34231906-34231906 on Build GRCh38
rs1008070149 Chr.15: 34231928-34231929 on Build GRCh38
rs528941290 Chr.15: 34231580-34231580 on Build GRCh38
rs1019461359 Chr.15: 34231964-34231964 on Build GRCh38
rs998862268 Chr.15: 34231654-34231654 on Build GRCh38
rs1028155199 Chr.15: 34231875-34231875 on Build GRCh38
rs567320325 Chr.15: 34231750-34231750 on Build GRCh38
rs774464841 Chr.15: 34232063-34232063 on Build GRCh38
rs907464298 Chr.15: 34231815-34231817 on Build GRCh38
rs953979177 Chr.15: 34231702-34231702 on Build GRCh38
rs886051044 Chr.15: 34231842-34231842 on Build GRCh38
rs573637671 Chr.15: 34232052-34232052 on Build GRCh38
rs953390564 Chr.15: 34231977-34231977 on Build GRCh38
rs770449723 Chr.15: 34231591-34231592 on Build GRCh38
rs987165220 Chr.15: 34232003-34232003 on Build GRCh38
rs200579522 Chr.15: 34231588-34231588 on Build GRCh38
rs1031729898 Chr.15: 34231661-34231661 on Build GRCh38
rs748784706 Chr.15: 34231588-34231589 on Build GRCh38
rs1045726068 Chr.15: 34231614-34231614 on Build GRCh38
rs202190008 Chr.15: 34232041-34232041 on Build GRCh38
rs184308650 Chr.15: 34231629-34231629 on Build GRCh38
rs553152328 Chr.15: 34231783-34231783 on Build GRCh38
rs143206625 Chr.15: 34231790-34231790 on Build GRCh38
rs566482685 Chr.15: 34231577-34231577 on Build GRCh38
rs140618820 Chr.15: 34232046-34232047 on Build GRCh38
rs539231349 Chr.15: 34231796-34231796 on Build GRCh38
rs557915731 Chr.15: 34231812-34231812 on Build GRCh38
rs886051041 Chr.15: 34231588-34231592 on Build GRCh38
rs919654507 Chr.15: 34231756-34231757 on Build GRCh38
rs531092709 Chr.15: 34231735-34231735 on Build GRCh38
rs998831174 Chr.15: 34231815-34231815 on Build GRCh38
rs748613902 Chr.15: 34231590-34231592 on Build GRCh38
rs923093610 Chr.15: 34231650-34231650 on Build GRCh38
rs921627230 Chr.15: 34231749-34231749 on Build GRCh38
rs774175991 Chr.15: 34231957-34231957 on Build GRCh38
rs1026638952 Chr.15: 34232051-34232051 on Build GRCh38
rs1055957108 Chr.15: 34231795-34231795 on Build GRCh38
rs963195233 Chr.15: 34231885-34231885 on Build GRCh38
rs541035283 Chr.15: 34232078-34232078 on Build GRCh38
rs745350561 Chr.15: 34231744-34231744 on Build GRCh38
rs1011389401 Chr.15: 34231676-34231676 on Build GRCh38
rs61577073 Chr.15: 34231917-34231918 on Build GRCh38
rs894503535 Chr.15: 34231806-34231806 on Build GRCh38
rs894470909 Chr.15: 34231682-34231682 on Build GRCh38
rs551973352 Chr.15: 34231641-34231641 on Build GRCh38
rs768245113 Chr.15: 34231746-34231746 on Build GRCh38
rs377402786 Chr.15: 34231587-34231592 on Build GRCh38
rs148811864 Chr.15: 34231577-34231584 on Build GRCh38
rs555476017 Chr.15: 34231683-34231683 on Build GRCh38
rs569942900 Chr.15: 34231672-34231672 on Build GRCh38
rs967190979 Chr.15: 34232068-34232068 on Build GRCh38
rs371222354 Chr.15: 34231739-34231739 on Build GRCh38
rs199512316 Chr.15: 34231798-34231798 on Build GRCh38
rs886051045 Chr.15: 34232066-34232066 on Build GRCh38
rs904695178 Chr.15: 34231792-34231792 on Build GRCh38
rs987161537 Chr.15: 34231710-34231710 on Build GRCh38
rs956591688 Chr.15: 34231615-34231615 on Build GRCh38
rs1036701595 Chr.15: 34231575-34231575 on Build GRCh38
rs983317329 Chr.15: 34231738-34231738 on Build GRCh38
rs945754240 Chr.15: 34231787-34231787 on Build GRCh38
rs912482725 Chr.15: 34231731-34231731 on Build GRCh38
rs886051043 Chr.15: 34231793-34231793 on Build GRCh38
rs973308980 Chr.15: 34231747-34231747 on Build GRCh38

View Full Product Details