Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD11
Entrez Gene ID
22984
Gene Name
programmed cell death 11
Gene Aliases
ALG-4, ALG4, NFBP, RRP5
Chromosome Location
Chr.10: 103396615 - 103446295 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014976.1 NP_055791.1 5 - 6 650 56
XM_005269647.3 XP_005269704.1 5 - 6 690 56
XM_011539538.2 XP_011537840.1 5 - 6 638 56
XM_011539539.2 XP_011537841.1 5 - 6 638 56
XM_011539540.1 XP_011537842.1 4 - 5 504 56
GenBank mRNA AK294971.1 - 5 - 6 679 56
AK307418.1 - 5 - 6 663 56
BC049838.1 - 5 - 6 651 56
BC064486.1 - 5 - 6 643 56
BC080560.1 - 5 - 6 643 56
BC111040.1 - 5 - 6 605 56
D80007.1 - 5 - 6 603 56

View Full Product Details