The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173474.1 | NP_001166945.1 | 1 - 2 | 232 | 82 |
NM_004192.3 | NP_004183.2 | 1 - 2 | 232 | 82 | |
XM_005274434.3 | XP_005274491.1 | 2 - 3 | 338 | 82 | |
GenBank mRNA | AK001090.1 | - | 1 - 2 | 128 | 82 |
AK090498.1 | - | 1 - 2 | 184 | 82 | |
AK225770.1 | - | 1 - 2 | 154 | 82 | |
AK297805.1 | - | 1 - 2 | 130 | 82 | |
BC002508.2 | - | 1 - 2 | 118 | 82 | |
BC010089.2 | - | 1 - 2 | 116 | 82 | |
DA005122.1 | - | 1 - 2 | 232 | 82 | |
DA301876.1 | - | 1 - 2 | 187 | 82 |